Assay Details
Gene Symbol: WASF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003931.2 | NP_003922.1 | 2 - 3 | 708 | 107 | |
XM_011536235.1 | XP_011534537.1 | 2 - 3 | 666 | 107 | ||
GenBank mRNA | D87459.1 | - | 1 - 2 | 113 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D87459.1, NM_003931.2, XM_011536235.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
