Assay Details
Gene Symbol: UBE2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004223.4 | NP_004214.1 | 2 - 3 | 218 | 86 | |
NM_198183.2 | NP_937826.1 | 2 - 3 | 577 | 86 | ||
GenBank mRNA | AF031141.1 | - | 2 - 3 | 119 | 86 | |
AF061736.1 | - | 2 - 3 | 169 | 86 | ||
AK093462.1 | - | 2 - 3 | 531 | 86 | ||
AK226165.1 | - | 2 - 3 | 272 | 86 | ||
AK315755.1 | - | 2 - 3 | 137 | 86 | ||
BC032491.1 | - | 2 - 3 | 218 | 5246109 | 86 | |
CR457126.1 | - | 2 - 3 | 122 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001795.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_198183.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein ubiquitination
modification-dependent protein catabolic process
translesion synthesis
ISG15-protein conjugation
negative regulation of type I interferon production
cellular protein metabolic process
ubiquitin-protein transferase activity
protein binding
ISG15 transferase activity
ubiquitin binding
cellular protein modification process
protein ubiquitination
modification-dependent protein catabolic process
translesion synthesis
ISG15-protein conjugation
negative regulation of type I interferon production
cellular protein metabolic process
ubiquitin-protein transferase activity
protein binding
ISG15 transferase activity
ubiquitin binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11020858.hg.1 | ENST00000287156.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11020858.hg.1 | ENST00000340573.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11020858.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11020858.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11020858.hg.1 | TC11001795.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11020860.hg.1 | ENST00000287156.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11020860.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11020860.hg.1 | TC11001795.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182439.hg.1 | TC1100010897.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182438.hg.1 | TC1100010897.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182445.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182445.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182445.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182445.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182445.hg.1 | TC1100010897.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182446.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182446.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182446.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182446.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100182446.hg.1 | TC1100010897.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000287156.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000340573.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000526659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000527022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | ENST00000528275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_004223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010897.hg.1 | NM_198183.2 | Refseq |
