Assay Details
| Gene Symbol: TGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004245.3 | NP_004236.1 | 2 - 3 | 202 | 109 | |
| GenBank mRNA | AF035961.1 | - | 2 - 3 | 202 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Differentiation-specific factors modulate epidermal CYP1-4 gene expression in human skin in response to retinoic acid and classic aryl hydrocarbon receptor ligands.
- Authors
- Du L, Neis MM, Ladd PA, Keeney DS
- Journal
- J Pharmacol Exp Ther - Page No 1162 (Fri Dec 01 00:00:00 EST 2006)
- Volume (Year)
- 319 (2006)
- Product Usage
- -

TGM5
Genomic Map