Assay Details
| Gene Symbol: NCAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004540.3 | NP_004531.2 | 17 - 18 | 2651 | 68 | |
| XM_017028353.1 | XP_016883842.1 | 18 - 19 | 2791 | 68 | ||
| XM_017028357.1 | XP_016883846.1 | 17 - 18 | 2526 | 68 | ||
| GenBank mRNA | AK299054.1 | - | 14 - 15 | 2164 | 68 | |
| BC052946.1 | - | 17 - 18 | 2651 | 5315109 | 68 | |
| U75330.1 | - | 17 - 18 | 2471 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000060.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21000309.hg.1 | ENST00000400546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000309.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000309.hg.1 | TC21000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000308.hg.1 | ENST00000284894.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000308.hg.1 | ENST00000400546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000308.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000308.hg.1 | TC21000060.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | TC2100006714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145424.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | TC2100006714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100145423.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000284894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000400546.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000461281.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000484983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | ENST00000486367.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006714.hg.1 | NM_004540.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 172702 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 199050 | - | - |

NCAM2
Genomic Map