| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006375.hg.1 |
TC09000573.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006375.hg.1 |
ENST00000374198.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006375.hg.1 |
ENST00000374199.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006375.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09006375.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900156655.hg.1 |
TC0900008484.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374198.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000374199.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
ENST00000488937.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_001244926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008484.hg.1 |
NM_004697.4 |
Refseq |