Assay Details
| Gene Symbol: GGPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037277.1 | NP_001032354.1 | 2 - 3 | 152 | 107 | |
| GenBank mRNA | AB016043.1 | - | 2 - 3 | 237 | 107 | |
| AB019036.1 | - | 2 - 3 | 278 | 107 | ||
| AF057698.1 | - | 2 - 3 | 306 | 107 | ||
| AF125394.1 | - | 2 - 3 | 215 | 107 | ||
| AK025139.1 | - | 2 - 3 | 300 | 107 | ||
| BC005252.1 | - | 2 - 3 | 300 | 4051481 | 107 | |
| BC067768.1 | - | 2 - 3 | 240 | 5295932 | 107 | |
| BI462253.1 | - | 2 - 3 | 327 | 107 | ||
| BI520491.1 | - | 2 - 3 | 155 | 107 | ||
| CB216084.1 | - | 2 - 3 | 318 | 107 | ||
| DB473584.1 | - | 2 - 3 | 328 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001944.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB016043.1,AB019036.1,AF057698.1,AF125394.1,BC005252.1,BC067768.1,NM_001037277.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cholesterol biosynthetic process
isoprenoid metabolic process
geranyl diphosphate biosynthetic process
geranylgeranyl diphosphate biosynthetic process
farnesyl diphosphate biosynthetic process
dimethylallyltranstransferase activity
farnesyltranstransferase activity
geranyltranstransferase activity
protein binding
metal ion binding
cholesterol biosynthetic process
isoprenoid metabolic process
geranyl diphosphate biosynthetic process
geranylgeranyl diphosphate biosynthetic process
farnesyl diphosphate biosynthetic process
dimethylallyltranstransferase activity
farnesyltranstransferase activity
geranyltranstransferase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | TC01001944.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | ENST00000282841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | ENST00000358966.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | ENST00000476121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | ENST00000488594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030051.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000254724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188812.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188812.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188812.hg.1 | TC0100012055.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188830.hg.1 | TC0100012055.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000254724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 130606 | - | - |

GGPS1
Genomic Map