Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000260363.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000352331.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000395392.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000537891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000558303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000558585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000559279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000559689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
ENST00000560125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
NM_004856.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
NM_138555.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005432.hg.1 |
TC15000638.hg.1 |
HTA TC |