Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK314163.1 | - | 1 - 2 | 92 | 80 | |
AW175781.1 | - | 1 - 2 | 80 | |||
BC005242.1 | - | 1 - 2 | 63 | 3997566 | 80 | |
BC011020.1 | - | 1 - 2 | 69 | 4287267 | 80 | |
BC029395.1 | - | 1 - 2 | 65 | 80 | ||
BC031017.1 | - | 1 - 2 | 64 | 4670801 | 80 | |
X98263.1 | - | 1 - 2 | 75 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK314163.1,BC005242.1,BC011020.1,BC029395.1,BC031017.1,X98263.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013243.hg.1 | ENST00000604049.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185570.hg.1 | TC1600011007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013243.hg.1 | ENST00000604049.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 217047 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 226998 | - | - |
