| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038382.hg.1 |
TC03002173.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038382.hg.1 |
ENST00000296350.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038382.hg.1 |
ENST00000296351.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038382.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038382.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000469783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000473501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000489445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013733.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000296350.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000296351.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000439320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
ENST00000491399.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
NM_005929.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
NM_033316.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202636.hg.1 |
TC0300013733.hg.1 |
Clariom D TC |