| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014985.hg.1 |
TC01000947.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014985.hg.1 |
ENST00000369851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014985.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014984.hg.1 |
TC01000947.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014984.hg.1 |
ENST00000369851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014984.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014981.hg.1 |
TC01000947.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014981.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014982.hg.1 |
TC01000947.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014982.hg.1 |
ENST00000369851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014982.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014983.hg.1 |
TC01000947.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014983.hg.1 |
ENST00000369851.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014983.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238542.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238542.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238542.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100238545.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238545.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238545.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100238546.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238546.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238546.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100238543.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238543.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238543.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100238544.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238544.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238544.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100238541.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238541.hg.1 |
NM_006496.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100238541.hg.1 |
TC0100018257.hg.1 |
Clariom D TC |