Assay Details
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006768.4 | NP_006759.3 | 1 - 2 | 289 | 113 | |
XM_011538789.2 | XP_011537091.1 | 1 - 2 | 208 | 113 | ||
XM_017019993.1 | XP_016875482.1 | 1 - 2 | 279 | 113 | ||
GenBank mRNA | AB208878.1 | - | 1 - 2 | 244 | 113 | |
AF035620.1 | - | 1 - 2 | 224 | 113 | ||
AK290379.1 | - | 1 - 2 | 229 | 113 | ||
AY332222.1 | - | 1 - 2 | 224 | 113 | ||
BC136698.1 | - | 1 - 2 | 224 | 9020688 | 113 | |
BC136699.1 | - | 1 - 2 | 224 | 9020689 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001974.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011538789.2,XM_017019993.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
Ras protein signal transduction
negative regulation of signal transduction
protein ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
protein binding
nuclear localization sequence binding
zinc ion binding
ligase activity
identical protein binding
ubiquitin binding
ubiquitin protein ligase activity
MAPK cascade
Ras protein signal transduction
negative regulation of signal transduction
protein ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
protein binding
nuclear localization sequence binding
zinc ion binding
ligase activity
identical protein binding
ubiquitin binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12025821.hg.1 | ENST00000419234.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025821.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025821.hg.1 | TC12001974.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12025818.hg.1 | ENST00000327551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025818.hg.1 | ENST00000419234.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025818.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025818.hg.1 | TC12001974.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12025822.hg.1 | ENST00000419234.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025822.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025822.hg.1 | TC12001974.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192852.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192852.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192852.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192852.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192852.hg.1 | TC1200011946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200192854.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192854.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192854.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192854.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192854.hg.1 | TC1200011946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200192853.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192853.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192853.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192853.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192853.hg.1 | TC1200011946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200192849.hg.1 | TC1200011946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000327551.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000419234.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | ENST00000547043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011946.hg.1 | NM_006768.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 216175 | - | - |
