Assay Details
Gene Symbol: PROSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007198.3 | NP_009129.1 | 1 - 2 | 178 | 94 | |
XM_017013001.1 | XP_016868490.1 | 1 - 2 | 1008 | 94 | ||
GenBank mRNA | AK291135.1 | - | 1 - 2 | 116 | 94 | |
AL136616.1 | - | 1 - 2 | 158 | 94 | ||
BC012334.1 | - | 1 - 2 | 168 | 3546307 | 94 | |
CR533532.1 | - | 1 - 2 | 101 | 94 | ||
DA086552.1 | - | 1 - 2 | 178 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000278.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08003850.hg.1 | TC08000278.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003850.hg.1 | ENST00000328195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003850.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003847.hg.1 | TC08000278.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003847.hg.1 | ENST00000328195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003847.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003846.hg.1 | TC08000278.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003846.hg.1 | ENST00000328195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003846.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000521631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000521631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003848.hg.1 | TC08000278.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08003848.hg.1 | ENST00000328195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003848.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150192.hg.1 | TC0800007309.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150194.hg.1 | TC0800007309.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000521631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000521631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000522808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523521.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007309.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150189.hg.1 | TC0800007309.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000328195.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000518036.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000520073.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523187.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523358.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | ENST00000523994.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | NM_007198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150191.hg.1 | TC0800007309.hg.1 | Clariom D TC |
