Assay Details
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012413.3 | NP_036545.1 | 2 - 3 | 391 | 62 | |
GenBank mRNA | AK290605.1 | - | 2 - 3 | 373 | 62 | |
BC020456.1 | - | 2 - 3 | 292 | 62 | ||
BC036721.1 | - | 2 - 3 | 270 | 5224222 | 62 | |
BC047756.2 | - | 2 - 3 | 368 | 6058335 | 62 | |
BI755122.1 | - | 2 - 3 | 426 | 62 | ||
X67731.1 | - | 1 - 2 | 73 | 62 | ||
X71125.1 | - | 2 - 3 | 280 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000237.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02003536.hg.1 | TC02000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003536.hg.1 | ENST00000338415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003536.hg.1 | ENST00000537448.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003536.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003532.hg.1 | TC02000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003532.hg.1 | ENST00000338415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003532.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003537.hg.1 | TC02000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003537.hg.1 | ENST00000338415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003537.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003540.hg.1 | TC02000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003540.hg.1 | ENST00000338415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003540.hg.1 | ENST00000537448.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003540.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02003533.hg.1 | TC02000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02003533.hg.1 | ENST00000338415.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02003533.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150272.hg.1 | TC0200007261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150275.hg.1 | TC0200007261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | ENST00000480050.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150279.hg.1 | TC0200007261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | ENST00000404976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150271.hg.1 | TC0200007261.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | ENST00000338415.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | ENST00000470075.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | NM_012413.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200150276.hg.1 | TC0200007261.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 152127 | - | - |
