Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021003.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021003.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021003.hg.1 |
TC19001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021004.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021004.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021004.hg.1 |
TC19001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021002.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021002.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021002.hg.1 |
TC19001540.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188483.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188483.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188483.hg.1 |
TC1900010706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188483.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188483.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010706.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188484.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188484.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188484.hg.1 |
TC1900010706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188484.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188484.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188485.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188485.hg.1 |
NM_013376.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188485.hg.1 |
TC1900010706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188485.hg.1 |
ENST00000357949.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188485.hg.1 |
NM_013376.3 |
Refseq |