Assay Details
Gene Symbol: LINC01558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026773.1 | 2 - 3 | 401 | 79 | ||
GenBank mRNA | AB016901.1 | - | 2 - 3 | 413 | 79 | |
BC027853.1 | - | 2 - 3 | 424 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002318.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05246129 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 347066 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 256081 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 95878 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 662773 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 140588 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 195836 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 31425 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P70814 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3233499 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4766 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4767 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 24553 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 5723 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55029 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18736 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P31934101 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3233499 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P70814 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P70814 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3233499 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00321728 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P70814 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P70813 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P70814 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8130985 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130814 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130816 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130983 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130815 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123406 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130987 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130984 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130812 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130813 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8130986 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8130811 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8130982 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8123405 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 207698_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1555095_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11760048_at | - | - |
Affymetrix Human Genome U219 Array | 11760048_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06029094.hg.1 | TC06002318.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06029094.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06029096.hg.1 | TC06002318.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06029096.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06029096.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600196067.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196067.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600196067.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196067.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600196067.hg.1 | TC0600013942.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600196071.hg.1 | TC0600013942.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000366822.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | ENST00000495520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013942.hg.1 | NR_026773.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1555095_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 207698_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 207698_at | - | - |
Human Genome U133 Plus 2.0 Array | 1555095_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw591615 | - | - |
Human Genome U95 Set | 31651_at | - | - |
Human Genome U95 Set | 31651_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555095_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207698_at9 | - | - |
Human Genome U95 Set | 63705_f_at | - | - |
Human Genome U95 Set | 63705_PM_f_at | - | - |
Human Genome U95 Set | 63906_f_at | - | - |
Human Genome U95 Set | 63906_PM_f_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030829 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030829 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790492 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1790492 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790492 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726152 | NR_026773.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726152 | BC027853.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1726152 | NR_026773.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1726152 | BC027853.1 | human_transcriptome |
