Assay Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301132.1 | NP_001288061.1 | 1 - 2 | 245 | 66 | |
| XM_011521369.2 | XP_011519671.1 | 1 - 2 | 96 | 66 | ||
| GenBank mRNA | AB023162.1 | - | 1 - 2 | 245 | 66 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB023162.1,NM_001301132.1,XM_011521369.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148440.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148440.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148440.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148440.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148440.hg.1 | TC1500006956.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148444.hg.1 | TC1500006956.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000416165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000560846.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | ENST00000561464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_001301132.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NM_014952.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006956.hg.1 | NR_125372.1 | Refseq |

BAHD1
Genomic Map