Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015344.2 | NP_056159.2 | 3 - 4 | 393 | 89 | |
| GenBank mRNA | AF063605.1 | - | 3 - 4 | 370 | 89 | |
| AK075363.1 | - | 3 - 4 | 337 | 89 | ||
| AY277595.1 | - | 3 - 4 | 365 | 89 | ||
| AY358674.1 | - | 3 - 4 | 365 | 89 | ||
| BC000642.2 | - | 3 - 4 | 376 | 3344487 | 89 | |
| CR541735.1 | - | 2 - 3 | 286 | 89 | ||
| CR541760.1 | - | 2 - 3 | 286 | 89 | ||
| DA258969.1 | - | 3 - 4 | 394 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08002578.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08003362.hg.1 | TC08002578.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003362.hg.1 | ENST00000321250.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003362.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003363.hg.1 | TC08002578.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003363.hg.1 | ENST00000321250.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003363.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003361.hg.1 | TC08002578.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003361.hg.1 | ENST00000321250.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003361.hg.1 | ENST00000518192.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08003361.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000442880.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000519466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000520682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | ENST00000523116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_001128208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012287.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000518001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185095.hg.1 | TC0800012287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | ENST00000518192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185096.hg.1 | TC0800012287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800185097.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185097.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185097.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185097.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185097.hg.1 | TC0800012287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800185098.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185098.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185098.hg.1 | ENST00000321250.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800185098.hg.1 | NM_015344.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800185098.hg.1 | TC0800012287.hg.1 | Clariom D TC |
Citations (1)
Transcriptional profiling in coronary artery disease: indications for novel markers of coronary collateralization.
- Authors
- Chittenden TW, Sherman JA, Xiong F, Hall AE, Lanahan AA, Taylor JM, Duan H, Pearlman JD, Moore JH, Schwartz SM, Simons M
- Journal
- Circulation - Page No 1811 (Tue Oct 24 00:00:00 EDT 2006)
- Volume (Year)
- 114 (2006)
- Product Usage
- -

LEPROTL1
Genomic Map