Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
TC01000375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
ENST00000335514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
ENST00000497986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005798.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152820.hg.1 |
TC0100007550.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000468425.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007550.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000335514.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000465645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
ENST00000497986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_016311.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_178190.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
NM_178191.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152819.hg.1 |
TC0100007550.hg.1 |
Clariom D TC |