Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005783.2 | NP_001005783.2 | 4 - 5 | 643 | 84 | |
| NM_016527.3 | NP_057611.1 | 3 - 4 | 423 | 84 | ||
| XM_011541559.1 | XP_011539861.1 | 2 - 3 | 296 | 84 | ||
| XM_011541560.1 | XP_011539862.1 | 2 - 3 | 296 | 84 | ||
| XM_011541561.1 | XP_011539863.1 | 2 - 3 | 296 | 84 | ||
| XM_017001421.1 | XP_016856910.1 | 4 - 5 | 979 | 84 | ||
| GenBank mRNA | AF231917.1 | - | 3 - 4 | 361 | 84 | |
| AK298289.1 | - | 4 - 5 | 643 | 84 | ||
| AY513277.1 | - | 3 - 4 | 84 | |||
| BC020863.1 | - | 4 - 5 | 610 | 4557798 | 84 | |
| BG428999.1 | - | 3 - 4 | 419 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001037.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid oxidation
cellular lipid metabolic process
(S)-2-hydroxy-acid oxidase activity
receptor binding
FMN binding
very-long-chain-(S)-2-hydroxy-acid oxidase activity
long-chain-(S)-2-hydroxy-long-chain-acid oxidase activity
medium-chain-(S)-2-hydroxy-acid oxidase activity
fatty acid oxidation
cellular lipid metabolic process
(S)-2-hydroxy-acid oxidase activity
receptor binding
FMN binding
very-long-chain-(S)-2-hydroxy-acid oxidase activity
long-chain-(S)-2-hydroxy-long-chain-acid oxidase activity
medium-chain-(S)-2-hydroxy-acid oxidase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01016446.hg.1 | TC01001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016446.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016446.hg.1 | ENST00000361035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016446.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016446.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016447.hg.1 | TC01001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016447.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016447.hg.1 | ENST00000361035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016447.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016447.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169143.hg.1 | TC0100009608.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169142.hg.1 | TC0100009608.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000325945.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000361035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000457318.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000482991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | ENST00000622548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_001005783.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009608.hg.1 | NM_016527.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 129462 | - | - |

HAO2
Genomic Map