Assay Details
| Gene Symbol: RAB8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016530.2 | NP_057614.1 | 3 - 4 | 340 | 113 | |
| XM_017022312.1 | XP_016877801.1 | 4 - 5 | 395 | 113 | ||
| GenBank mRNA | AB038995.1 | - | 3 - 4 | 335 | 113 | |
| AK001111.1 | - | 3 - 4 | 263 | 113 | ||
| AK297598.1 | - | 3 - 4 | 263 | 113 | ||
| AL833365.1 | - | 3 - 4 | 253 | 113 | ||
| BC020654.1 | - | 3 - 4 | 252 | 4701429 | 113 | |
| BG721755.1 | - | 3 - 4 | 404 | 113 | ||
| U66624.1 | - | 1 - 2 | 47 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000581.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017022312.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
vesicle docking involved in exocytosis
small GTPase mediated signal transduction
protein secretion
antigen processing and presentation
positive regulation of cell projection organization
adherens junction organization
cilium assembly
protein import into peroxisome membrane
positive regulation of corticotropin secretion
GTPase activity
receptor binding
protein binding
GTP binding
GDP binding
TPR domain binding
vesicle docking involved in exocytosis
small GTPase mediated signal transduction
protein secretion
antigen processing and presentation
positive regulation of cell projection organization
adherens junction organization
cilium assembly
protein import into peroxisome membrane
positive regulation of corticotropin secretion
GTPase activity
receptor binding
protein binding
GTP binding
GDP binding
TPR domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15004654.hg.1 | ENST00000321437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004654.hg.1 | ENST00000448330.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004654.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004654.hg.1 | TC15000581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004655.hg.1 | ENST00000321437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004655.hg.1 | ENST00000448330.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004655.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004655.hg.1 | TC15000581.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500153257.hg.1 | TC1500007518.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500153256.hg.1 | TC1500007518.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000321437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558119.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000558990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | ENST00000559927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007518.hg.1 | NM_016530.2 | Refseq |

RAB8B
Genomic Map