| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020144.hg.1 |
ENST00000328550.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020144.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020144.hg.1 |
ENST00000451802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020144.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020144.hg.1 |
TC19001485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020153.hg.1 |
ENST00000328550.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020153.hg.1 |
ENST00000451802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020153.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020153.hg.1 |
TC19001485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020154.hg.1 |
ENST00000451802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020154.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020154.hg.1 |
TC19001485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020155.hg.1 |
ENST00000328550.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020155.hg.1 |
ENST00000451802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020155.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19020155.hg.1 |
TC19001485.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
TC1900010574.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900186861.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000328550.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000358744.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000451802.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590390.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000590751.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
ENST00000593133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_001290314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010574.hg.1 |
NM_016536.4 |
Refseq |