Assay Details
Gene Symbol: SNRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100594.1 | NP_001094064.1 | 2 - 3 | 857 | 87 | |
NM_017719.4 | NP_060189.3 | 3 - 4 | 919 | 87 | ||
XM_005265245.3 | XP_005265302.1 | 4 - 5 | 969 | 87 | ||
GenBank mRNA | AK000231.1 | - | 1 - 2 | 679 | 87 | |
AK291360.1 | - | 4 - 5 | 970 | 87 | ||
AK314376.1 | - | 2 - 3 | 742 | 87 | ||
BC071567.1 | - | 2 - 3 | 831 | 4375623 | 87 | |
D43636.2 | - | 1 - 2 | 591 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000229.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | TC03000229.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | ENST00000296088.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | ENST00000429705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | ENST00000454177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03004642.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300150669.hg.1 | TC0300007189.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000296088.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000429705.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000437827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000454177.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000462810.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000468628.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000481892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | ENST00000484791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_001100594.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007189.hg.1 | NM_017719.4 | Refseq |
