Assay Details
| Gene Symbol: LAMTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017907.2 | NP_060377.1 | 1 - 2 | 209 | 68 | |
| GenBank mRNA | AF289583.1 | - | 1 - 2 | 100 | 68 | |
| AK000632.1 | - | 1 - 2 | 98 | 68 | ||
| BC001706.1 | - | 1 - 2 | 71 | 3544000 | 68 | |
| BX381569.2 | - | 1 - 2 | 209 | 68 | ||
| CR457247.1 | - | 1 - 2 | 46 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002053.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of receptor recycling
endosome organization
lysosome organization
cell cycle arrest
regulation of cholesterol esterification
regulation of cholesterol efflux
cell growth
macroautophagy
positive regulation of TOR signaling
lysosome localization
endosome localization
cellular protein localization
cholesterol homeostasis
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cholesterol import
cellular response to amino acid stimulus
guanyl-nucleotide exchange factor activity
protein binding
protein complex scaffold
regulation of receptor recycling
endosome organization
lysosome organization
cell cycle arrest
regulation of cholesterol esterification
regulation of cholesterol efflux
cell growth
macroautophagy
positive regulation of TOR signaling
lysosome localization
endosome localization
cellular protein localization
cholesterol homeostasis
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cholesterol import
cellular response to amino acid stimulus
guanyl-nucleotide exchange factor activity
protein binding
protein complex scaffold
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | ENST00000278671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | ENST00000535107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | ENST00000545249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024495.hg.1 | TC11002053.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | ENST00000278671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | ENST00000535107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | ENST00000539797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | ENST00000545249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024493.hg.1 | TC11002053.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024499.hg.1 | ENST00000278671.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024499.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11024499.hg.1 | TC11002053.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188881.hg.1 | TC1100011543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011543.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000535872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000539797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000544594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188871.hg.1 | TC1100011543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100188883.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188883.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188883.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188883.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188883.hg.1 | TC1100011543.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000278671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000535107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000538404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000541403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | ENST00000545249.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | NM_017907.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100188875.hg.1 | TC1100011543.hg.1 | Clariom D TC |

LAMTOR1
Genomic Map