Assay Details
Gene Symbol: LRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018214.4 | NP_060684.4 | 1 - 2 | 434 | 67 | |
XM_011514726.2 | XP_011513028.1 | 2 - 3 | 798 | 67 | ||
XM_017010997.1 | XP_016866486.1 | 1 - 2 | 726 | 67 | ||
GenBank mRNA | AF332199.1 | - | 1 - 2 | 381 | 67 | |
AF359380.1 | - | 1 - 2 | 157 | 67 | ||
AK001637.1 | - | 1 - 2 | 138 | 67 | ||
AK021896.1 | - | 2 - 3 | 798 | 67 | ||
AK292248.1 | - | 1 - 2 | 279 | 67 | ||
BC003193.1 | - | 1 - 2 | 434 | 3504090 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000668.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001637.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06007990.hg.1 | TC06000668.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007990.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007990.hg.1 | ENST00000370888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007990.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007988.hg.1 | TC06000668.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007988.hg.1 | ENST00000370888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007988.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06007989.hg.1 | TC06000668.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06007989.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007989.hg.1 | ENST00000370888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06007989.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157945.hg.1 | TC0600008303.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157946.hg.1 | TC0600008303.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | ENST00000490222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008303.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157943.hg.1 | TC0600008303.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000370882.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000370888.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | ENST00000487251.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | NM_018214.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157944.hg.1 | TC0600008303.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 223624 | - | - |
