Assay Details
| Gene Symbol: H2AFJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK001765.1 | - | 1 - 2 | 417 | 89 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |

H2AFJ
Genomic Map