Assay Details
Gene Symbol: TCP10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK001794.1 | - | 1 - 2 | 1305 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000300258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000334165.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000472557.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000491828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | ENST00000582575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008558.hg.1 | NM_144659.6 | Refseq |
