Assay Details
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002154.1 | - | 1 - 2 | 200 | 74 | |
BC117434.1 | - | 1 - 2 | 85 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077188 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257443 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257413 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077183 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077173 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077126 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257426 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257425 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077170 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15717844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077149 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257470 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077104 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077191 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077095 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077112 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257412 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257414 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257454 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257501 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24257411 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04077184 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3567961 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3567967 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P371722 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P122210 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 14088 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P202039 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P124167 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P124167 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15056 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3567961 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3567967 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P122210 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P371722 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P202039 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P124167 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17198851 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17198828 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P202039 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P124167 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P371722 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P122210 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8113004 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8113005 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8113003 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220828_PM_s_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 217342_PM_x_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 215187_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 217335_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000445770.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | ENST00000611331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011387.hg.1 | NR_130914.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 217342_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 215187_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 217335_at | - | - |
Human Genome U133 Plus 2.0 Array | 217335_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 217342_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 220828_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 215187_at | - | - |
Human Genome U133 Plus 2.0 Array | 220828_s_at | - | - |
Human Genome U133A 2.0 Array | 215187_at | - | - |
Human Genome U133A 2.0 Array | 217342_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 220828_s_at | - | - |
Human Genome U133A 2.0 Array | 215187_PM_at | - | - |
Human Genome U133A 2.0 Array | 220828_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 217335_PM_at | - | - |
Human Genome U133A 2.0 Array | 217342_x_at | - | - |
Human Genome U133A 2.0 Array | 217335_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_018382_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw516645 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw584046 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw523097 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw556091 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220828_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217342_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217335_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215187_at11 | - | - |
Human Genome U133 Set | 220828_PM_s_at | - | - |
Human Genome U133 Set | 217335_at | - | - |
Human Genome U133 Set | 215187_PM_at | - | - |
Human Genome U133 Set | 217335_PM_at | - | - |
Human Genome U133 Set | 215187_at | - | - |
Human Genome U133 Set | 217342_x_at | - | - |
Human Genome U133 Set | 220828_s_at | - | - |
Human Genome U133 Set | 217342_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 137527 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1695562 | AK024354.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1695562 | AK023417.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1695562 | AK002104.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1695562 | AK002154.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790753 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_1695562 | AK024354.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1695562 | AK023417.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1695562 | AK002104.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1695562 | AK002154.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922980-S | AK024354.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922980-S | AK023417.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922980-S | AK002104.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_8922980-S | AK002154.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_15574-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005270601 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005270601 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1790753 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790753 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 360168 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_15574 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1695562 | AK024354.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1695562 | AK023417.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1695562 | AK002104.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1695562 | AK002154.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695562 | AK024354.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695562 | AK023417.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695562 | AK002104.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1695562 | AK002154.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1790753 | - | - |
