Assay Details
| Gene Symbol: NCBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024828.1 | XP_016880317.1 | 1 - 2 | 89 | 87 | |
| XM_017024829.1 | XP_016880318.1 | 1 - 2 | 89 | 87 | ||
| GenBank mRNA | AJ277841.1 | - | 1 - 2 | 94 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ277841.1, XM_017024828.1, XM_017024829.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000571336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000572988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000572988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000576523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000576523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000389005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000571336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000572988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000572988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000574911.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000575815.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000576523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000576523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | ENST00000577169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009490.hg.1 | NM_001114118.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 194092 | - | - |

NCBP3
Genomic Map