Assay Details
| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042478.1 | NP_001035943.1 | 1 - 2 | 413 | 105 | |
| NM_018836.3 | NP_061324.1 | 1 - 2 | 413 | 105 | ||
| XM_011541786.2 | XP_011540088.1 | 1 - 2 | 683 | 105 | ||
| GenBank mRNA | AF175409.1 | - | 1 - 2 | 725 | 105 | |
| AY282806.1 | - | 1 - 2 | 413 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000080.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF175409.1,AY282806.1,NM_001042478.1,NM_018836.3,XM_011541786.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01001238.hg.1 | TC01000080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001238.hg.1 | ENST00000378190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001238.hg.1 | ENST00000378191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001238.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001238.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | TC01000080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | ENST00000378190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | ENST00000378191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001241.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001239.hg.1 | TC01000080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001239.hg.1 | ENST00000378190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001239.hg.1 | ENST00000378191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001239.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001239.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001242.hg.1 | TC01000080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001242.hg.1 | ENST00000378190.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001242.hg.1 | ENST00000378191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001242.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01001242.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146013.hg.1 | TC0100006650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146018.hg.1 | TC0100006650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | ENST00000466761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006650.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146015.hg.1 | TC0100006650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | ENST00000378190.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | ENST00000378191.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | NM_001042478.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | NM_018836.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100146019.hg.1 | TC0100006650.hg.1 | Clariom D TC |

AJAP1
Genomic Map