Assay Details
| Gene Symbol: PPP4R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020958.2 | NP_066009.2 | 3 - 4 | 450 | 97 | |
| XM_017021530.1 | XP_016877019.1 | 3 - 4 | 408 | 97 | ||
| GenBank mRNA | BC002650.2 | - | 3 - 4 | 424 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000585.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017021530.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14007219.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007219.hg.1 | ENST00000555690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007219.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007219.hg.1 | TC14000585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157166.hg.1 | TC1400008058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 205087 | - | - |

PPP4R4
Genomic Map