Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008826.hg.1 |
ENST00000246024.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008826.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008826.hg.1 |
TC20000611.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
TC2000008345.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158819.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000246024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000462384.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000527925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
ENST00000530935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008345.hg.1 |
NM_021156.3 |
Refseq |