Assay Details
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022157.3 | NP_071440.1 | 1 - 2 | 413 | 60 | |
GenBank mRNA | AF272035.1 | - | 1 - 2 | 241 | 60 | |
AF323609.1 | - | 1 - 2 | 236 | 60 | ||
AK315791.1 | - | 1 - 2 | 338 | 60 | ||
BC016668.1 | - | 1 - 2 | 258 | 4342448 | 60 | |
BG717489.1 | - | 1 - 2 | 421 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01006351.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
cell cycle arrest
small GTPase mediated signal transduction
RNA splicing
regulation of autophagy
cell growth
macroautophagy
regulation of TOR signaling
positive regulation of TOR signaling
cellular response to amino acid starvation
cellular protein localization
response to amino acid
cellular response to amino acid stimulus
regulation of TORC1 signaling
magnesium ion binding
GTPase activity
protein binding
GTP binding
GDP binding
protein heterodimerization activity
transcription, DNA-templated
apoptotic process
cell cycle arrest
small GTPase mediated signal transduction
RNA splicing
regulation of autophagy
cell growth
macroautophagy
regulation of TOR signaling
positive regulation of TOR signaling
cellular response to amino acid starvation
cellular protein localization
response to amino acid
cellular response to amino acid stimulus
regulation of TORC1 signaling
magnesium ion binding
GTPase activity
protein binding
GTP binding
GDP binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01039428.hg.1 | TC01006351.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039428.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039428.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039429.hg.1 | TC01006351.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039429.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039429.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039426.hg.1 | TC01006351.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039426.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039426.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01039430.hg.1 | TC01006351.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01039430.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01039430.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241824.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241824.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241824.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241824.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241824.hg.1 | TC0100018433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241823.hg.1 | TC0100018433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241825.hg.1 | TC0100018433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241827.hg.1 | TC0100018433.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000373001.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000474456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000493015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | ENST00000496778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_001271851.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018433.hg.1 | NM_022157.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175356 | - | - |
