Assay Details
| Gene Symbol: TNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022748.11 | NP_073585.8 | 23 - 24 | 3900 | 60 | |
| XM_011515476.2 | XP_011513778.1 | 23 - 24 | 4781 | 60 | ||
| XM_011515477.2 | XP_011513779.1 | 23 - 24 | 3933 | 60 | ||
| XM_011515478.2 | XP_011513780.1 | 23 - 24 | 4009 | 60 | ||
| XM_011515479.2 | XP_011513781.1 | 20 - 21 | 6085 | 60 | ||
| XM_011515480.2 | XP_011513782.1 | 23 - 24 | 4031 | 60 | ||
| XM_011515481.2 | XP_011513783.1 | 21 - 22 | 3697 | 60 | ||
| XM_011515482.2 | XP_011513784.1 | 23 - 24 | 5386 | 60 | ||
| XM_011515483.2 | XP_011513785.1 | 20 - 21 | 3930 | 60 | ||
| XM_011515484.2 | XP_011513786.1 | 8 - 9 | 3460 | 60 | ||
| XM_017012537.1 | XP_016868026.1 | 22 - 23 | 3870 | 60 | ||
| XM_017012538.1 | XP_016868027.1 | 23 - 24 | 4177 | 60 | ||
| XM_017012539.1 | XP_016868028.1 | 22 - 23 | 5283 | 60 | ||
| GenBank mRNA | AB062750.1 | - | 7 - 8 | 1298 | 60 | |
| AF378756.1 | - | 11 - 12 | 2914 | 60 | ||
| AF417489.1 | - | 18 - 19 | 3551 | 60 | ||
| AK023794.1 | - | 1 - 2 | 41 | 60 | ||
| AL833845.1 | - | 7 - 8 | 1864 | 60 | ||
| BC137133.1 | - | 19 - 20 | 3580 | 9021130 | 60 | |
| BC137134.1 | - | 19 - 20 | 3580 | 9021131 | 60 | |
| CR749644.1 | - | 23 - 24 | 3849 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001363.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023794.1 transcripts.
Set Membership: |
|

TNS3
Genomic Map