Assay Details
| Gene Symbol: CAMKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024766.4 | NP_079042.1 | 7 - 8 | 729 | 90 | |
| XM_011533111.2 | XP_011531413.1 | 7 - 8 | 695 | 90 | ||
| XM_011533113.2 | XP_011531415.1 | 5 - 6 | 405 | 90 | ||
| XM_017004982.1 | XP_016860471.1 | 5 - 6 | 2832 | 90 | ||
| XM_017004983.1 | XP_016860472.1 | 6 - 7 | 944 | 90 | ||
| GenBank mRNA | AK027104.1 | - | 7 - 8 | 669 | 90 | |
More Information
Set Membership: |
|

CAMKMT
Genomic Map