Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000567369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000567369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000623838.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009638.hg.1 |
ENST00000623838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000563613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000567369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000567369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000623838.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009638.hg.1 |
ENST00000623838.1 |
Ensembl |