Assay Details
| Gene Symbol: SERPINB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030666.3 | NP_109591.1 | 2 - 3 | 413 | 108 | |
| NR_073112.1 | 2 - 3 | 413 | 108 | |||
| XM_011514333.1 | XP_011512635.1 | 3 - 4 | 636 | 108 | ||
| XM_011514334.2 | XP_011512636.1 | 2 - 3 | 547 | 108 | ||
| GenBank mRNA | AK223370.1 | - | 2 - 3 | 226 | 108 | |
| AK291327.1 | - | 2 - 3 | 226 | 108 | ||
| AK304641.1 | - | 2 - 3 | 258 | 108 | ||
| AK312973.1 | - | 2 - 3 | 224 | 108 | ||
| BC009015.1 | - | 2 - 3 | 211 | 3456154 | 108 | |
| BT006928.1 | - | 1 - 2 | 167 | 108 | ||
| BU942473.1 | - | 2 - 3 | 228 | 108 | ||
| EU794624.1 | - | 1 - 2 | 167 | 108 | ||
| GQ472232.1 | - | 2 - 3 | 211 | 108 | ||
| M93056.1 | - | 2 - 3 | 215 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001217.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304641.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | TC06001217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | ENST00000476896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | ENST00000537185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | ENST00000542771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014481.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | TC06001217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | ENST00000476896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | ENST00000537185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | ENST00000542771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06014482.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600200148.hg.1 | TC0600014217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000380739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000460260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000468511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000476896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | ENST00000490094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NM_030666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073111.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014217.hg.1 | NR_073112.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124046 | - | - |

SERPINB1
Genomic Map