Assay Details
| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL136611.1 | - | 7 - 8 | 1752 | 113 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000331314.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000413408.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000429498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000433235.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000436897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000447706.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000452560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | ENST00000453944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016893.hg.1 | NM_001031725.5 | Refseq |

DDX59
Genomic Map