Assay Details
| Gene Symbol: REG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159352.1 | NP_001152824.1 | 3 - 4 | 505 | 58 | |
| NM_001159353.1 | NP_001152825.1 | 2 - 3 | 331 | 58 | ||
| NM_032044.3 | NP_114433.1 | 2 - 3 | 331 | 58 | ||
| GenBank mRNA | AF254415.1 | - | 2 - 3 | 246 | 58 | |
| AF345934.1 | - | 2 - 3 | 199 | 58 | ||
| AY007243.1 | - | 2 - 3 | 187 | 58 | ||
| AY126670.1 | - | 3 - 4 | 505 | 58 | ||
| AY126671.1 | - | 2 - 3 | 331 | 58 | ||
| AY126672.1 | - | 2 - 3 | 331 | 58 | ||
| BC017089.2 | - | 2 - 3 | 203 | 3850358 | 58 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003051.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047515.hg.1 | TC01003051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047515.hg.1 | ENST00000354219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01047515.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215561.hg.1 | TC0100015477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000256585.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000369401.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | ENST00000530654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_001159353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015477.hg.1 | NM_032044.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215564.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215564.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215564.hg.1 | ENST00000354219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100215564.hg.1 | NM_001159352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100215564.hg.1 | TC0100015477.hg.1 | Clariom D TC |

REG4
Genomic Map