Assay Details
| Gene Symbol: IKBKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190720.2 | NP_001177649.1 | 7 - 8 | 758 | 61 | |
| NM_001242778.1 | NP_001229707.1 | 7 - 8 | 786 | 61 | ||
| NM_001556.2 | NP_001547.1 | 8 - 9 | 881 | 61 | ||
| NR_033818.1 | 8 - 9 | 937 | 61 | |||
| NR_033819.1 | 8 - 9 | 881 | 61 | |||
| NR_040009.1 | 8 - 9 | 881 | 61 | |||
| XM_005273490.2 | XP_005273547.1 | 8 - 9 | 881 | 61 | ||
| XM_005273491.4 | XP_005273548.1 | 7 - 8 | 781 | 61 | ||
| XM_005273492.3 | XP_005273549.1 | 8 - 9 | 881 | 61 | ||
| XM_005273493.3 | XP_005273550.1 | 5 - 6 | 577 | 61 | ||
| XM_005273494.2 | XP_005273551.1 | 8 - 9 | 881 | 61 | ||
| XM_011544517.2 | XP_011542819.1 | 8 - 9 | 879 | 61 | ||
| XM_011544519.2 | XP_011542821.1 | 7 - 8 | 749 | 61 | ||
| XM_011544520.2 | XP_011542822.1 | 5 - 6 | 596 | 61 | ||
| XM_011544522.2 | XP_011542824.1 | 5 - 6 | 557 | 61 | ||
| XM_017013396.1 | XP_016868885.1 | 7 - 8 | 834 | 61 | ||
| GenBank mRNA | AB209090.1 | - | 7 - 8 | 699 | 61 | |
| AF029684.1 | - | 7 - 8 | 695 | 61 | ||
| AF031416.1 | - | 7 - 8 | 695 | 61 | ||
| AF080158.1 | - | 8 - 9 | 838 | 61 | ||
| AK290952.1 | - | 8 - 9 | 747 | 61 | ||
| AK298241.1 | - | 5 - 6 | 445 | 61 | ||
| AK302723.1 | - | 7 - 8 | 742 | 61 | ||
| AK303294.1 | - | 7 - 8 | 770 | 61 | ||
| AK303528.1 | - | 7 - 8 | 774 | 61 | ||
| AK316418.1 | - | 7 - 8 | 742 | 61 | ||
| BC108694.1 | - | 8 - 9 | 793 | 5784717 | 61 | |
| BC144080.1 | - | 7 - 8 | 725 | 61 | ||
| BX648165.1 | - | 7 - 8 | 1333 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000325.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011544522.2,XM_017013396.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
NAK is recruited to the TNFR1 complex in a TNFalpha-dependent manner and mediates the production of RANTES: identification of endogenous TNFR-interacting proteins by a proteomic approach.
- Authors
- Kuai J, Wooters J, Hall JP, Rao VR, Nickbarg E, Li B, Chatterjee-Kishore M, Qiu Y, Lin LL
- Journal
- J Biol Chem - Page No 53266 (Fri Dec 17 00:00:00 EST 2004)
- Volume (Year)
- 279 (2004)
- Product Usage
- -

IKBKB
Genomic Map