| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15011775.hg.1 |
ENST00000290650.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15011775.hg.1 |
ENST00000382177.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15011775.hg.1 |
ENST00000546274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15011775.hg.1 |
NM_174916.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15011775.hg.1 |
TC15001271.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000290650.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000546274.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000564540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000569066.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000569243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
NM_174916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000290650.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000546274.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000564540.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000569066.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
ENST00000569243.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
NM_174916.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167011.hg.1 |
TC1500009204.hg.1 |
Clariom D TC |