| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000241261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000430881.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000494851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
NM_003810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
NR_033994.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000241261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000430881.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
ENST00000494851.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
NM_003810.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
NR_033994.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300198241.hg.1 |
TC0300013146.hg.1 |
Clariom D TC |