Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158479.hg.1 |
TC0200008452.hg.1 |
Clariom D TC |