Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004185.4 | NP_004176.2 | 2 - 3 | 1128 | 78 | |
| GenBank mRNA | AB045116.1 | - | 2 - 3 | 251 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000989.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte differentiation
lens development in camera-type eye
male gonad development
cellular response to starvation
Wnt signaling pathway
forebrain regionalization
neuron differentiation
cell fate commitment
canonical Wnt signaling pathway
lung induction
mesenchymal-epithelial cell signaling
iris morphogenesis
cornea development in camera-type eye
hematopoietic stem cell proliferation
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of canonical Wnt signaling pathway
frizzled binding
chondrocyte differentiation
lens development in camera-type eye
male gonad development
cellular response to starvation
Wnt signaling pathway
forebrain regionalization
neuron differentiation
cell fate commitment
canonical Wnt signaling pathway
lung induction
mesenchymal-epithelial cell signaling
iris morphogenesis
cornea development in camera-type eye
hematopoietic stem cell proliferation
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of canonical Wnt signaling pathway
frizzled binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01015725.hg.1 | TC01000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015725.hg.1 | ENST00000369686.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015725.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015724.hg.1 | TC01000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015724.hg.1 | ENST00000256640.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015724.hg.1 | ENST00000369686.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015724.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167959.hg.1 | TC0100009442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167954.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167954.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167954.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167954.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167954.hg.1 | TC0100009442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167951.hg.1 | TC0100009442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167952.hg.1 | TC0100009442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167953.hg.1 | TC0100009442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000256640.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369684.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000369686.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | ENST00000478360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_001291880.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_004185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009442.hg.1 | NM_024494.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 128778 | - | - |

WNT2B
Genomic Map