Assay Details
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004397.5 | NP_004388.2 | 1 - 2 | 169 | 128 | |
XM_011542644.1 | XP_011540946.1 | 1 - 2 | 169 | 128 | ||
XM_011542645.1 | XP_011540947.1 | 1 - 2 | 169 | 128 | ||
GenBank mRNA | BC039826.1 | - | 1 - 2 | 53 | 128 | |
DA993470.1 | - | 1 - 2 | 169 | 128 | ||
Z11685.1 | - | 1 - 2 | 69 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002353.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC039826.1,NM_004397.5,XM_011542644.1,XM_011542645.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
RNA secondary structure unwinding
viral RNA genome packaging
stem cell population maintenance
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
cell-cell adhesion
RNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
protein domain specific binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
regulation of translation
RNA secondary structure unwinding
viral RNA genome packaging
stem cell population maintenance
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
negative regulation of neuron differentiation
cell-cell adhesion
RNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
protein domain specific binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000525082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000526070.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000529162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000531971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000533239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000534980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000617381.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | ENST00000620157.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_001257191.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012495.hg.1 | NM_004397.5 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 107193 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 233158 | - | - |
