Assay Details
Gene Symbol: KIF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194313.2 | NP_919289.2 | 2 | 373 | 62 | |
XM_011517861.1 | XP_011516163.1 | 3 | 594 | 62 | ||
XM_011517863.2 | XP_011516165.1 | 2 | 1683 | 62 | ||
XM_017014697.1 | XP_016870186.1 | 2 | 687 | 62 | ||
XM_017014698.1 | XP_016870187.1 | 2 | 558 | 62 | ||
GenBank mRNA | AK310014.1 | - | 3 | 594 | 62 | |
BU660279.1 | - | 2 | 377 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001031.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017014698.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | TC09001031.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | ENST00000345050.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | ENST00000379174.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | ENST00000402558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | ENST00000420188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013521.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | TC09001031.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | ENST00000345050.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | ENST00000379174.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | ENST00000402558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | ENST00000420188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013522.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09013525.hg.1 | TC09001031.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09013525.hg.1 | ENST00000345050.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013525.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013525.hg.1 | ENST00000420188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09013525.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000410494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167785.hg.1 | TC0900009892.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900167788.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167788.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167788.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167788.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167788.hg.1 | TC0900009892.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167787.hg.1 | TC0900009892.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000410494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000379166.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000379174.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | ENST00000402558.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | NM_194313.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900167786.hg.1 | TC0900009892.hg.1 | Clariom D TC |
