Assay Details
| Gene Symbol: PCDHA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018904.2 | NP_061727.1 | 1 - 2 | 2401 | 67 | |
| GenBank mRNA | AB208924.1 | - | 1 - 2 | 2462 | 67 | |
| AF152309.1 | - | 1 - 2 | 2401 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000737.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | TC05000737.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000253807.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000289269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000289272.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000307360.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000394633.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000398631.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000398640.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000409494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000504120.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000506939.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000512229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000522353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000525929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000526136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000527624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000529310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000529619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000529859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000530339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000531613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | ENST00000532602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018899.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018900.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018902.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018903.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018904.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018905.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018906.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018907.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018908.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018910.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_018911.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_031411.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_031849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_031857.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05010714.hg.1 | NM_031860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000253807.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000289269.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000289272.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000307360.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000394633.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000398631.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000398640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000409494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000504120.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000506939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000512229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000522353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000525929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000526136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000527624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000530339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000531613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000532602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000617408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018898.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018899.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018900.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018903.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018904.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018905.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018906.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018907.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018908.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018910.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031411.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000253807.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000289269.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000289272.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000307360.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000394633.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000398631.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000398640.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000409494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000504120.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000506939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000512229.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000522353.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000525929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000526136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000527624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529619.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000529859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000530339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000531613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000532602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | ENST00000617408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018898.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018899.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018900.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018902.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018903.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018904.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018905.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018906.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018907.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018908.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018910.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_018911.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031411.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031857.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | NM_031860.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500161635.hg.1 | TC0500008871.hg.1 | Clariom D TC |

PCDHA13
Genomic Map