Assay Details
| Gene Symbol: CDC42SE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020239.3 | NP_064624.1 | 1 - 2 | 174 | 60 | |
| GenBank mRNA | AF187845.2 | - | 1 - 2 | 99 | 60 | |
| AF286592.1 | - | 1 - 2 | 99 | 60 | ||
| BC041604.1 | - | 1 - 2 | 136 | 5473010 | 60 | |
| BP340339.1 | - | 1 - 2 | 174 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF187845.2, AF286592.1, BC041604.1, NM_020239.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000357235.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000470278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000483763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000491825.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000492796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | ENST00000540998.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_001038707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018486.hg.1 | NM_020239.3 | Refseq |

CDC42SE1
Genomic Map