Assay Details
| Gene Symbol: FBXW4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033408.1 | 1 | 398 | 98 | ||
| GenBank mRNA | AF174606.1 | - | 1 | 398 | 98 | |
| AK024483.1 | - | 1 | 2711 | 98 | ||
| AK314175.1 | - | 1 | 261 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000130.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF174606.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P252603 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 17186 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P252603 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P252603 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 4400011 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 4400011 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8071714 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8071713 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 214623_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11760550_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11760551_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11760551_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11760550_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002335.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002335.hg.1 | TC22000130.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008712.hg.1 | ENST00000471845.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008159.hg.1 | ENST00000467969.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007845.hg.1 | ENST00000464637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009892.hg.1 | ENST00000581481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010743.hg.1 | ENST00000625125.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009069.hg.1 | ENST00000570247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009069.hg.1 | ENST00000570247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009892.hg.1 | ENST00000581481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009069.hg.1 | ENST00000570247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009069.hg.1 | ENST00000570247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008712.hg.1 | ENST00000471845.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147118.hg.1 | TC2200006836.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147118.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147118.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147118.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147118.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007845.hg.1 | ENST00000464637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008159.hg.1 | ENST00000467969.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010743.hg.1 | ENST00000610674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010743.hg.1 | ENST00000625125.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147119.hg.1 | TC2200006836.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147119.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147119.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147119.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147119.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147120.hg.1 | TC2200006836.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147120.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147120.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147120.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147120.hg.1 | NR_033408.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 214623_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214623_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214623_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214623_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw513707 | - | - |
| Human Genome U95 Set | 517_at | - | - |
| Human Genome U95 Set | 517_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214623_at11 | - | - |
| Human Genome U95 Set | 76675_f_at | - | - |
| Human Genome U95 Set | 76675_PM_f_at | - | - |
| Human Genome U133 Set | 214623_at | - | - |
| Human Genome U133 Set | 214623_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020197 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 199316 | - | - |

FBXW4P1
Genomic Map