Assay Details
| Gene Symbol: USH1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK024943.1 | - | 1 | 201 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK024943.1 transcripts.
Set Membership: |
|

USH1C
Genomic Map