Assay Details
| Gene Symbol: KIF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB210045.1 | - | 29 | 4488 | 66 | |
| AB290174.1 | - | 29 | 4729 | 66 | ||
| AF063608.1 | - | 1 | 630 | 66 | ||
| BC051752.1 | - | 1 | 705 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KIF5A
Genomic Map